3-(3,4-Methylendioxy)phenylpropionsäure

Lieferant: FLUOROCHEM

Synonyme: 3,4-(Methylenedioxy)hydrocinnamic acid, 3-(1,3-Benzodioxyl-5-yl)propionic acid

018840-5G
FLUO018840-5GEA 192 EUR
FLUO018840-5G
3-(3,4-Methylendioxy)phenylpropionsäure
3-(3,4-Methylendioxy)phenylpropionsäure
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
MDL: MFCD00016909
CAS-Nummer: 2815-95-4
EINECS: 220-565-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR