Methyl-5-methylsalicylat

Lieferant: FLUOROCHEM

Synonyme: Methyl 2-hydroxy-5-methylbenzoate

018844-1G
FLUO018844-1GEA 47.7 EUR
FLUO018844-1G
Methyl-5-methylsalicylat
Methyl-5-methylsalicylat
Formel: C₉H₁₀O₃
Molecular Weight: 166,1700 g/mol
MDL: MFCD00016485
CAS-Nummer: 22717-57-3
EINECS: 245-173-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR