Methyl-(3-methoxyphenyl)acetat

Lieferant: FLUOROCHEM

Synonyme: Methyl 3-Methoxyphenylacetate

018845-10G
FLUO018845-10GEA 140 EUR
FLUO018845-10G
Methyl-(3-methoxyphenyl)acetat
Methyl-(3-methoxyphenyl)acetat
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,23 g/mol
MDL: MFCD00017205
CAS-Nummer: 18927-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR