5-Methyl-4-phenyl-o-anisidin

Lieferant: FLUOROCHEM

018853-1G
FLUO018853-1GEA 156 EUR
FLUO018853-1G
5-Methyl-4-phenyl-o-anisidin
5-Methyl-4-phenyl-o-anisidin
Formel: C₁₄H₁₅NO
MDL: MFCD00270117
CAS-Nummer: 206761-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR