1-(3-Methylphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM
Gefahr

018862-2G
FLUO018862-2GEA 0 EUR
FLUO018862-2G
1-(3-Methylphenyl)-2-thioharnstoff
1-(3-Methylphenyl)-2-thioharnstoff
Formel: (C₆H₄(CH₃))NC(S)NH₂
Molecular Weight: 166,25 g/mol
MDL: MFCD00060451
CAS-Nummer: 621-40-9
EINECS: 210-686-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR