6-Methylchinaldin
Lieferant: FLUOROCHEM
Synonyme:
2,6-Dimethylchinolin
018867-10G
FLUO018867-10GEA
221
EUR
FLUO018867-10G
6-Methylchinaldin
6-Methylchinaldin
Formel:
C₁₁H₁₁N Molecular Weight: 157,22 g/mol Siedepunkt: 266 °C (1013 hPa) Schmelzpunkt: 55…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006762 CAS-Nummer: 877-43-0 EINECS: 212-891-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...