4-(Methylsulfonyl)benzylchlorid

Lieferant: FLUOROCHEM

018869-5G
FLUO018869-5GEA 87.9 EUR
FLUO018869-5G
4-(Methylsulfonyl)benzylchlorid
4-(Methylsulfonyl)benzylchlorid
Formel: C₈H₉ClO₂S
MDL: MFCD00079774
CAS-Nummer: 40517-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR