2-(Methylthio)nicotinsäurehydrazid

Lieferant: FLUOROCHEM

018876-1G
FLUO018876-1GEA 84.8 EUR
FLUO018876-1G
2-(Methylthio)nicotinsäurehydrazid
2-(Methylthio)nicotinsäurehydrazid
Formel: C₇H₉N₃OS
CAS-Nummer: 232926-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR