1-Methyl-2-thioharnstoff

Lieferant: FLUOROCHEM

018883-25G
FLUO018883-25GEA 53.6 EUR
FLUO018883-25G
1-Methyl-2-thioharnstoff
1-Methyl-2-thioharnstoff
Formel: C₂H₆N₂S
Molecular Weight: 90,15 g/mol
Schmelzpunkt: 119 °C
MDL: MFCD00004938
CAS-Nummer: 598-52-7
EINECS: 209-936-6
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR