Methyl-o-toluat
Lieferant: FLUOROCHEM
Synonyme:
2-Tolylsäuremethylester, o-Toluylsäuremethylester, 2-Methylbenzoesäuremethylester, Methyl-2-methylbenzoat, 2-Toluylsäuremethylester
018884-25G
FLUO018884-25GEA
34.1
EUR
FLUO018884-25G
Methyl-o-toluat
Methyl-o-toluat
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Siedepunkt: 215 °C (1013 hPa) Schmelzpunkt: –50 °C Dichte: 1,079 g/cm³ (0 °C) |
MDL:
MFCD00008428 CAS-Nummer: 89-71-4 EINECS: 201-932-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...