2-Nitrobenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
2-Nitrobenzoic hydrazide
018899-5G
FLUO018899-5GEA
0
EUR
FLUO018899-5G
2-Nitrobenzhydrazid
2-Nitrobenzhydrazid
Formel:
C₇H₇N₃O₃ Molecular Weight: 181,15 g/mol |
MDL:
MFCD00007598 CAS-Nummer: 606-26-8 EINECS: 210-110-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...