3-Nitrobenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
3-Nitrobenzoic hydrazide
018900-10G
FLUO018900-10GEA
0
EUR
FLUO018900-10G
3-Nitrobenzhydrazid
3-Nitrobenzhydrazid
Formel:
C₇H₇N₃O₃ Molecular Weight: 181,15 g/mol |
MDL:
MFCD00017059 CAS-Nummer: 618-94-0 EINECS: 210-572-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...