4-Nitrobenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
4-Nitrobenzoic hydrazide
018901-10G
FLUO018901-10GEA
94.1
EUR
FLUO018901-10G
4-Nitrobenzhydrazid
4-Nitrobenzhydrazid
Formel:
C₇H₇N₃O₃ Molecular Weight: 181,15 g/mol |
MDL:
MFCD00007604 CAS-Nummer: 636-97-5 EINECS: 211-271-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...