4'-Nitrobenzo-15-krone-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecan)
Lieferant: FLUOROCHEM
Synonyme:
4-Nitrobenzo-15-crown-5
018902-1G
FLUO018902-1GEA
128
EUR
FLUO018902-1G
4'-Nitrobenzo-15-krone-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecan)
4'-Nitrobenzo-15-krone-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel:
C₁₄H₁₉NO₇ Molecular Weight: 313.30 g/mol |
MDL:
MFCD00060713 CAS-Nummer: 60835-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...