5-Nitroisatosäureanhydrid
Lieferant: FLUOROCHEM
Synonyme:
6-Nitro-2H-3,1-benzoxazine-2,4(1H)-dione
018904-5G
FLUO018904-5GEA
39.6
EUR
FLUO018904-5G
5-Nitroisatosäureanhydrid
5-Nitroisatosäureanhydrid
Formel:
C₈H₄N₂O₅ Molecular Weight: 208,13 g/mol |
MDL:
MFCD00016919 CAS-Nummer: 20829-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...