4-(4-Nitrophenoxy)anilin

Lieferant: FLUOROCHEM

018906-5G
FLUO018906-5GEA 159 EUR
FLUO018906-5G
4-(4-Nitrophenoxy)anilin
4-(4-Nitrophenoxy)anilin
Formel: C₁₂H₁₀N₂O₃
MDL: MFCD00060609
CAS-Nummer: 6149-33-3
EINECS: 228-159-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR