(3-Nitrophenyl)acetonitril
Lieferant: FLUOROCHEM
018907-5G
FLUO018907-5GEA
87.9
EUR
FLUO018907-5G
(3-Nitrophenyl)acetonitril
(3-Nitrophenyl)acetonitril
Formel:
C₈H₆N₂O₂ Siedepunkt: 145…150 °C (0,5 mmHg) Schmelzpunkt: 63 °C |
MDL:
MFCD00041249 CAS-Nummer: 621-50-1 EINECS: 210-689-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...