1-(4-Nitrophenyl)imidazol

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Nitrophenyl)-1H-imidazol

018909-1G
FLUO018909-1GEA 35.9 EUR
FLUO018909-1G
1-(4-Nitrophenyl)imidazol
1-(4-Nitrophenyl)imidazole
Formel: C₉H₇N₃O₂
Molecular Weight: 189,17 g/mol
MDL: MFCD00060491
CAS-Nummer: 2301-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR