4-(2-Nitrophenyl)-3-thiosemicarbazid

Lieferant: FLUOROCHEM

018916-1G
FLUO018916-1GEA 0 EUR
FLUO018916-1G
4-(2-Nitrophenyl)-3-thiosemicarbazid
4-(2-Nitrophenyl)-3-thiosemicarbazid
Formel: C₇H₈N₄O₂S
MDL: MFCD00060568
CAS-Nummer: 73305-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR