1-Methyl-2-(trichloracetyl)pyrrol

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trichloro-1-(1-methyl-1H-pyrrol-2-yl)ethanone

018917-25G
FLUO018917-25GEA 174 EUR
FLUO018917-25G
1-Methyl-2-(trichloracetyl)pyrrol
1-Methyl-2-(trichloracetyl)pyrrol
Formel: C₇H₆Cl₃NO
Molecular Weight: 226,4876 g/mol
MDL: MFCD00052521
CAS-Nummer: 21898-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR