1-(3-Nitrophenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
|
Gefahr
|
018921-1G
FLUO018921-1GEA
82.1
EUR
FLUO018921-1G
1-(3-Nitrophenyl)-2-thioharnstoff
1-(3-Nitrophenyl)-2-thioharnstoff
Formel:
(C₆H₄(NO₂))NC(S)NH₂ Molecular Weight: 197.22 g/mol |
MDL:
MFCD00060541 CAS-Nummer: 709-72-8 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...