1-(4-Nitrophenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM
Gefahr

Synonyme: N-(4-Nitrophenyl)thioharnstoff, 4-Nitrophenylthioharnstoff

018922-5G
FLUO018922-5GEA 0 EUR
FLUO018922-5G
1-(4-Nitrophenyl)-2-thioharnstoff
1-(4-Nitrophenyl)-2-thioharnstoff
Formel: (C₆H₄(NO₂))NC(S)NH₂
Molecular Weight: 197,22 g/mol
Schmelzpunkt: 245 °C
MDL: MFCD00007304
CAS-Nummer: 3696-22-8
EINECS: 223-021-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR