3-(tert-Octylamino)propionitril

Lieferant: FLUOROCHEM

018929-2G
FLUO018929-2GEA 0 EUR
FLUO018929-2G
3-(tert-Octylamino)propionitril
3-(tert-Octylamino)propionitril
Formel: C₁₁H₂₂N₂
MDL: MFCD00060311
CAS-Nummer: 86375-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR