5-Phenoxy-o-anisidin
Lieferant: FLUOROCHEM
018944-1G
FLUO018944-1GEA
224
EUR
FLUO018944-1G
5-Phenoxy-o-anisidin
5-Phenoxy-o-anisidin
Formel:
C₁₃H₁₃NO₂ |
MDL:
MFCD00270120 CAS-Nummer: 116289-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...