4-Phenoxybenzoesäure

Lieferant: FLUOROCHEM
Achtung

018947-10G
FLUO018947-10GEA 45.9 EUR
FLUO018947-10G
4-Phenoxybenzoesäure
4-Phenoxybenzoesäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,22 g/mol
MDL: MFCD00002539
CAS-Nummer: 2215-77-2
EINECS: 218-682-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR