3-Phenoxybenzylchlorid
Lieferant: FLUOROCHEM
018948-1G
018948-5G
018948-25G
FLUO018948-1GEA
0
EUR
FLUO018948-1G
FLUO018948-5G
FLUO018948-25G
3-Phenoxybenzylchlorid
3-Phenoxybenzylchlorid
Formel:
C₁₃H₁₁ClO Siedepunkt: 123 °C (0,3 mmHg) Dichte: 1,16 g/cm³ (20 °C) |
MDL:
MFCD00040866 CAS-Nummer: 53874-66-1 EINECS: 258-831-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...