Phenylacetamid
Lieferant: FLUOROCHEM
Synonyme:
2-Phenylacetamide
018952-25G
FLUO018952-25GEA
36.9
EUR
FLUO018952-25G
Phenylacetamid
Phenylacetamid
Formel:
C₈H₉NO Molecular Weight: 135,17 g/mol |
MDL:
MFCD00059193 CAS-Nummer: 103-81-1 EINECS: 203-147-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...