3-Phenyl-o-anisidin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: 6-Methoxy-[1,1'-biphenyl]-3-amine hydrochloride

018953-1G
FLUO018953-1GEA 246 EUR
FLUO018953-1G
3-Phenyl-o-anisidin Hydrochlorid
3-Phenyl-o-anisidin Hydrochlorid
Formel: C₁₃H₁₄ClNO
Molecular Weight: 235,7100 g/mol
MDL: MFCD01630736
CAS-Nummer: 92028-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR