4-Phenyl-m-anisidin Hydrochlorid

Lieferant: FLUOROCHEM

018954-1G
FLUO018954-1GEA 222 EUR
FLUO018954-1G
4-Phenyl-m-anisidin Hydrochlorid
4-Phenyl-m-anisidin Hydrochlorid
Formel: C₁₃H₁₄ClNO
MDL: MFCD01569251
CAS-Nummer: 206761-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR