6-Phenyl-m-anisidin Hydrochlorid

Lieferant: FLUOROCHEM

018956-1G
FLUO018956-1GEA 200 EUR
FLUO018956-1G
6-Phenyl-m-anisidin Hydrochlorid
6-Phenyl-m-anisidin Hydrochlorid
Formel: C₁₃H₁₄ClNO
MDL: MFCD00270121
CAS-Nummer: 107624-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR