2-Phenylbenzhydrazid

Lieferant: FLUOROCHEM

018958-1G
FLUO018958-1GEA 162 EUR
FLUO018958-1G
2-Phenylbenzhydrazid
2-Phenylbenzhydrazid
Formel: C₁₃H₁₂N₂O
MDL: MFCD00270122
CAS-Nummer: 154660-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR