Phenyl-4-hydroxybenzoat
Lieferant: FLUOROCHEM
018964-10G
FLUO018964-10GEA
82.1
EUR
FLUO018964-10G
Phenyl-4-hydroxybenzoat
Phenyl-4-hydroxybenzoat
Formel:
C₁₃H₁₀O₃ |
MDL:
MFCD00016467 CAS-Nummer: 17696-62-7 EINECS: 241-698-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...