5-Phenyl-o-toluidin

Lieferant: FLUOROCHEM

Synonyme: 4-Methyl-[1,1'-biphenyl]-3-amine, 3-Amino-4-methylbiphenyl

018972-1G
FLUO018972-1GEA 167 EUR
FLUO018972-1G
5-Phenyl-o-toluidin
5-Phenyl-o-toluidin
Formel: C₁₃H₁₃N
Molecular Weight: 183,26 g/mol
MDL: MFCD00270126
CAS-Nummer: 80938-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR