1-Propyl-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: N-(n-Propyl)thiourea, 1-Propylthiourea

018984-5G
FLUO018984-5GEA 296 EUR
FLUO018984-5G
1-Propyl-2-thioharnstoff
1-Propyl-2-thioharnstoff
Formel: C₄H₁₀N₂S
Molecular Weight: 118,2 g/mol
MDL: MFCD00041195
CAS-Nummer: 927-67-3
EINECS: 213-158-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR