N-(2-Pyridyl)thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: (2-Pyridyl)thiourea, 1-(2-Pyridyl)-2-thiourea, 1-(Pyridin-2-yl)thiourea

018986-10G
FLUO018986-10GEA 0 EUR
FLUO018986-10G
N-(2-Pyridyl)thioharnstoff
N-(2-Pyridyl)thioharnstoff
Formel: C₆H₇N₃S
Molecular Weight: 153.21 g/mol
MDL: MFCD00041227
CAS-Nummer: 14294-11-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR