(2,3,5,6-Tetramethylphenyl)acetonitril

Lieferant: FLUOROCHEM

018996-1G
FLUO018996-1GEA 149 EUR
FLUO018996-1G
(2,3,5,6-Tetramethylphenyl)acetonitril
(2,3,5,6-Tetramethylphenyl)acetonitril
Formel: C₁₂H₁₅N
Molecular Weight: 173.26 g/mol
MDL: MFCD00060297
CAS-Nummer: 14611-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR