1,1,3,3-Tetramethyl-2-thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: Tetramethylthioharnstoff, Tetramethyl-2-thioharnstoff, N,N,N',N'-Tetramethylthioharnstoff

018997-25G
FLUO018997-25GEA 0 EUR
FLUO018997-25G
1,1,3,3-Tetramethyl-2-thioharnstoff
1,1,3,3-Tetramethyl-2-thioharnstoff
Formel: C₅H₁₂N₂S
Molecular Weight: 132,23 g/mol
Siedepunkt: 245 °C (1013 hPa)
Schmelzpunkt: 74…76 °C
MDL: MFCD00008324
CAS-Nummer: 2782-91-4
EINECS: 220-488-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR