m-Toluylsäurehydrazid
Lieferant: FLUOROCHEM
Synonyme:
m-Toluic acid hydrazide, 3-Methylbenzoic hydrazide
019001-10G
FLUO019001-10GEA
222
EUR
FLUO019001-10G
m-Toluylsäurehydrazid
m-Toluylsäurehydrazid
Formel:
C₈H₁₀N₂O Molecular Weight: 150.18 g/mol |
MDL:
MFCD00014760 CAS-Nummer: 13050-47-0 EINECS: 235-926-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...