o-Toluylsäurehydrazid

Lieferant: FLUOROCHEM

Synonyme: o-Toluic acid hydrazide, 2-Methylbenzohydrazide

019002-10G
FLUO019002-10GEA 0 EUR
FLUO019002-10G
o-Toluylsäurehydrazid
o-Toluylsäurehydrazid
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
MDL: MFCD00014756
CAS-Nummer: 7658-80-2
EINECS: 231-623-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR