p-Toluylsäurehydrazid

Lieferant: FLUOROCHEM

Synonyme: p-Toluoylhydrazine, 4-Methylbenzohydrazide, p-Toluic acid hydrazide, p-Toluic Hydrazide

019003-10G
FLUO019003-10GEA 0 EUR
FLUO019003-10G
p-Toluylsäurehydrazid
p-Toluylsäurehydrazid
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
MDL: MFCD00007607
CAS-Nummer: 3619-22-5
EINECS: 222-815-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR