1-(2,4,5-Trichlorphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

019007-1G
FLUO019007-1GEA 0 EUR
FLUO019007-1G
1-(2,4,5-Trichlorphenyl)-2-thioharnstoff
1-(2,4,5-Trichlorphenyl)-2-thioharnstoff
Formel: (C₆H₂Cl₃)NC(S)NH₂
Molecular Weight: 255,55 g/mol
MDL: MFCD00060431
CAS-Nummer: 90617-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR