1-(2,4,6-Trichlorphenyl)-2-thioharnstoff
Lieferant: FLUOROCHEM
Synonyme:
2,4,6-Trichlorphenylthioharnstoff, N-(2,4,6-Trichlorphenyl)thioharnstoff
019008-1G
FLUO019008-1GEA
67.1
EUR
FLUO019008-1G
1-(2,4,6-Trichlorphenyl)-2-thioharnstoff
1-(2,4,6-Trichlorphenyl)-2-thioharnstoff
Formel:
(C₆H₂Cl₃)NC(S)NH₂ Molecular Weight: 255,55 g/mol |
MDL:
MFCD00041153 CAS-Nummer: 31118-87-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...