2,3,4-Trichlortoluol

Lieferant: FLUOROCHEM

Synonyme: 1-Methyl-2,3,4-trichlorbenzol

019009-5G
FLUO019009-5GEA 0 EUR
FLUO019009-5G
2,3,4-Trichlortoluol
2,3,4-Trichlortoluol
Formel: C₇H₅Cl₃
Molecular Weight: 195,47 g/mol
MDL: MFCD00018098
CAS-Nummer: 7359-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR