2,4,6-Triisopropylbenzoesäure
Lieferant: FLUOROCHEM
019014-10G
FLUO019014-10GEA
146
EUR
FLUO019014-10G
2,4,6-Triisopropylbenzoesäure
2,4,6-Triisopropylbenzoesäure
Formel:
C₁₆H₂₄O₂ Molecular Weight: 248,37 g/mol |
MDL:
MFCD00015031 CAS-Nummer: 49623-71-4 EINECS: 256-400-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...