2,4,6-Trimethoxybenzylalkohol
Lieferant: FLUOROCHEM
019017-1G
FLUO019017-1GEA
128
EUR
FLUO019017-1G
2,4,6-Trimethoxybenzylalkohol
2,4,6-Trimethoxybenzylalkohol
Formel:
C₁₀H₁₄O₄ Molecular Weight: 198,22 g/mol |
MDL:
MFCD00060356 CAS-Nummer: 61040-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...