1-(3,4,5-Trimethoxyphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM
Gefahr

019021-1G
FLUO019021-1GEA 0 EUR
FLUO019021-1G
1-(3,4,5-Trimethoxyphenyl)-2-thioharnstoff
1-(3,4,5-Trimethoxyphenyl)-2-thioharnstoff
Formel: (C₆H₂(OCH₃)₃)NC(S)NH₂
Molecular Weight: 242.3 g/mol
MDL: MFCD00060447
CAS-Nummer: 59083-54-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR