1-(4-Chlorbenzyl)piperazin

Lieferant: FLUOROCHEM

019041-5G
FLUO019041-5GEA 0 EUR
FLUO019041-5G
1-(4-Chlorbenzyl)piperazin
1-(4-Chlorbenzyl)piperazin
Formel: C₁₁H₁₅ClN₂
Molecular Weight: 210,71 g/mol
MDL: MFCD00040791
CAS-Nummer: 23145-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR