1-(2-Pyridin-4-ylethyl)piperazin

Lieferant: FLUOROCHEM

Synonyme: 1-(2-(Pyridin-4-yl)ethyl)piperazine

019053-5G
FLUO019053-5GEA 312 EUR
FLUO019053-5G
1-(2-Pyridin-4-ylethyl)piperazin
1-(2-Pyridin-4-ylethyl)piperazin
Formel: C₁₁H₁₇N₃
Molecular Weight: 191,27 g/mol
MDL: MFCD03094584
CAS-Nummer: 53345-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR