1-(2-Pyridin-2-ylethyl)piperazin

Lieferant: FLUOROCHEM

019054-5G
FLUO019054-5GEA 0 EUR
FLUO019054-5G
1-(2-Pyridin-2-ylethyl)piperazin
1-(2-Pyridin-2-ylethyl)piperazin
Formel: C₁₁H₁₇N₃
MDL: MFCD01748645
CAS-Nummer: 53345-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR