4-Iod-1-tritylimidazol

Lieferant: FLUOROCHEM

Synonyme: 4-Iodo-1-trityl-1H-imidazole

019056-1G
FLUO019056-1GEA 42 EUR
FLUO019056-1G
4-Iod-1-tritylimidazol
4-Iod-1-tritylimidazol
Formel: C₂₂H₁₇IN₂
Molecular Weight: 436,2900 g/mol
MDL: MFCD02179542
CAS-Nummer: 96797-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR